Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Acetylpyrazine 99 FCC FG5KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W3126065KGK
Encompass
2-Acetylpyrazine has been identified as one of the volatile flavor constituents in popcorn wheat and rye bread crust.
Catalog No. 50-290-7124