Learn More
Medchemexpress LLC Ethylene dimethanesulfonate | 4672-49-5 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY129524100MG
Encompass_Preferred
Ethylene dimethanesulfonate is a mild alkylating, non-volatile methanesulfonic diester of ethylene glycol. It exhibits selective pro-apoptotic effects on LCs.
  • Mild alkylating agent
  • Non-volatile methanesulfonic diester
  • Selective pro-apoptotic effects on LCs
  • White to off-white solid appearance
  • Disrupts epididymal function (in vivo)
Catalog No. 50-003-39452