Learn More
Polysciences, Inc. Ethylene glycol diglycidyl ether (EGDGE)
SDP
Supplier:  Polysciences, Inc. 01479100
Encompass
Difunctional, aliphatic epoxy with usage in cross-linking, adhesives, coatings, and other biomedical or industrial applications.
Catalog No. NC1929723