Learn More
Medchemexpress LLC Ethylene glycol dimethacrylate | 97-90-5 | MFCD00008590 | 99.5% | 198.22 g/mol | C10H14O4 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW014940
Encompass_Preferred
Ethylene glycol dimethacrylate is a methacrylate diester monomer used primarily as a cross-linking agent in polymer, resin, and coating formulations. It is a liquid (colorless to light yellow) that promotes network formation to improve mechanical properties and stability of polymer systems. Due to its biological activity, it is intended for controlled research and material applications and should be handled with appropriate safety precautions.
  • Acts as a cross-linking agent for polymer networks.
  • Enhances mechanical strength and dimensional stability.
  • Compatible with resins, coatings, and adhesive formulations.
  • Liquid form facilitates incorporation into formulations.
  • Available at high purity for reproducible results.
  • Requires careful handling due to cytotoxic and genotoxic effects.
Catalog No. 50-003-02474