Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethylene glycol monohexadecyl ether | 2136-71-2 | MFCD00043350 | ≥97.0% | 286.49 g/mol | C18H38O2 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW250466100MG

Encompass_Preferred

Ethylene glycol monohexadecyl ether is a nonionic surfactant used as a biochemical assay reagent in research laboratories. It provides consistent performance in assay formulations and reagent preparations, and is supplied with characterization data for use in experimental workflows.

  • Nonionic surfactant for biochemical assays.
  • High purity: ≥97.0%.
  • Molecular weight: 286.49 g/mol; chemical formula: C18H38O2.
  • Supplier-reported CAS number: 2136-71-2.
  • Storage stability: room temperature up to 3 years; in solvent -80°C (2 years), -20°C (1 year).

Catalog No. 50-002-81236


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.