Learn More
Medchemexpress LLC Ethylene glycol monohexadecyl ether | 2136-71-2 | MFCD00043350 | ≥97.0% | 286.49 g/mol | C18H38O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW25046650MG
Encompass_Preferred
Ethylene glycol monohexadecyl ether is a nonionic surfactant used as a solubilizing and detergent reagent in biochemical and membrane assays. It is intended for research use only, has molecular formula C18H38O2 and molecular weight ≈286.49 g/mol, and is supplied at high purity for reliable experimental results.
  • Nonionic surfactant suitable for solubilizing membrane proteins.
  • High purity (≥97.0%) for research applications.
  • Small-scale packaging for assay development and optimization.
  • Compatible with biochemical and membrane-based assay workflows.
  • Accompanied by safety data sheets in multiple languages.
Catalog No. 50-002-81820