Learn More
Medchemexpress LLC Ethylhexadecyldimeth 500g
SDP
Supplier:  Medchemexpress LLC HYW099576500G
Encompass_Preferred
Ethylhexadecyldimethylammonium (EHDA) bromide a surfactant has been used in a number of adsorptive separational methods such as the removal of nickel zinc and chromium ions Ethylhexadecyldimethylammonium (EHDA) bromide also can be used to prepare dye of staining intracellular ions[1][2]
Catalog No. 50-002-05440