Learn More
Medchemexpress LLC Ethylhexyl triazone | 88122-99-0 | 98.0% | 823.07 | 1 ML

Supplier: Medchemexpress LLC HY1096551G
Ethylhexyl triazone is an approved ultraviolet-B (UV-B) chemical filter for commercial sunscreens. It possesses a large molecular weight, which suggests it is unlikely to penetrate the skin, thereby reducing associated effects. It displays a broad absorption profile across the UV-B region.
- UV-B filter
- High molecular weight (823.07 g/mol) reduces skin penetration
- Broad absorption profile across the UV-B region
- Maximum absorption at approximately 311 nm (in dioxane) and 313 nm (in methanol)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.