Learn More
Cayman Chemical EthylLNIO hydrochloride
SDP
Supplier:  Cayman Chemical 1001208810 MG
Encompass_Preferred
A modestly selective NOS inhibitor with Ki values for inhibition of nNOS, eNOS, and iNOS of 5.3, 18, and 12 µM, respectively; however, does not show biologically significant selectivity for nNOS over eNOS, and instead favors iNOS (Ki/Km = 3.79, 5, and 0.96 µM, respectively)
Catalog No. 50-288-7869