Learn More
Medchemexpress LLC Ethylmalonic acid-d3 | 70907-93-6 | 98.0% | 135.13 | C5H5D3O4 | 10 MG

Supplier: Medchemexpress LLC HY34740S10MG
Ethylmalonic acid-d3 is the deuterium-labeled analogue of ethylmalonic acid used as an analytical standard and stable isotope tracer in metabolic research and mass spectrometry. Supplied as a white to off-white solid in milligram quantities, it is intended for research use only.
- Deuterium-labeled internal standard for mass spectrometry and quantification.
- High chemical purity: 98.0%.
- Molecular weight: 135.13 g/mol.
- Chemical formula: C5H5D3O4, d3 isotopologue suitable for tracer studies.
- Supplied in small milligram pack sizes for analytical applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.