Learn More
Medchemexpress LLC Ethylparaben-d4 | 1219795-53-5 | MFCD16039539 | 99.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0934S1MG
Encompass_Preferred
Ethylparaben-d4 is a deuterium-labeled Ethylparaben. It functions as a tracer and internal standard for quantitative analysis, primarily in drug development processes. The incorporation of stable heavy isotopes, such as deuterium, can influence the pharmacokinetic and metabolic profiles of drug molecules.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Contains stable heavy isotopes
  • Deuteration may affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-07883