Learn More
Medchemexpress LLC Ethynylferrocene | 1271-47-2 | MFCD03701501 | 97.0% | 210.05 | C12H10Fe | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW018008500MG
Encompass_Preferred
Ethynylferrocene is an organometallic building block (ferrocenylacetylene) used in click chemistry and organic synthesis for labeling, ligand construction, and cross-coupling reactions. It is a stable solid (C12H10Fe, MW 210.05) commonly handled under inert atmosphere and protected from light for storage and long-term stability.
  • Used as a click-chemistry reagent and synthetic building block.
  • Solid organometallic compound with a defined molecular formula and weight.
  • Suitable for labeling, ligand synthesis, and Sonogashira or azide-alkyne cycloaddition reactions.
  • Store protected from light under inert atmosphere and refrigerated to maintain stability.
  • Supplied in small-scale quantities appropriate for research and development work.
Catalog No. 50-003-02042