Learn More
Sigma Organic Chemistry Ethynyltrimethylsilane | 25G | 1066-54-2 | MFCD00008569
Supplier: Sigma Organic Chemistry 21817025G
CAS #: 1066-54-2
MDL #: MFCD00008569
Purity: >98 %
UNSPSC Code: 12352100
Molecular Weight: 98.22
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.