Learn More
Medchemexpress LLC Etidocaine hydrochloride | 36637-19-1 | 99.9% | 1 MG

Supplier: Medchemexpress LLC HYB2080A1MG
Etidocaine hydrochloride, also known as EDC hydrochloride, is a long aminoamide local agent developed to suppress or relieve pain. This product is intended for research use only and is not suitable for patient use.
- Purity of 99.93%
- Molecular weight of 312.88
- Formula: C17H29ClN2O
- Appears as a white to off-white solid
- Can be stored as powder at -20°C for 3 years or 4°C for 2 years
- When in solvent, store at -80°C for 6 months or -20°C for 1 month
- Soluble in DMSO at 62.5 mg/mL (199.76 mM)
- Exhibits biological activity in vitro by increasing human fibroblast survival
- In vivo studies show no post-injection neurologic deficit in adult Swiss Webster male mice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.