Learn More
Medchemexpress LLC Etidronic acid, 60% in water | 2809-21-4 | 60% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB0302
Encompass_Preferred
Etidronic acid, 60% in water, is an analytical standard intended for research and analytical applications. It is an orally and intravenously active bisphosphonate that inhibits bone resorption and reduces arterial calcification. It can also be used for osteoporosis research and exhibits anticancer activity, acting as a chelating agent for heavy metal removal in water.
  • Analytical standard for research and analytical applications
  • Inhibits bone resorption
  • Reduces arterial calcification
  • Used for osteoporosis research
  • Exhibits anticancer activity
  • Acts as a chelating agent for heavy metal removal in water
Catalog No. 50-004-06523