Learn More
Medchemexpress LLC Etidronic acid (etidronate) | 2809-21-4 | 98.0% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB030250G
Encompass_Preferred
Etidronic acid (Etidronate) is an orally and intravenously active bisphosphonate that inhibits resorption of bone, reduces arterial calcification, and can be used for osteoporosis research. It also exhibits anticancer activity and acts as a chelating agent for removing heavy metals in water.
  • Inhibits resorption of bone
  • Reduces arterial calcification
  • Can be used for osteoporosis research
  • Exhibits anticancer activity
  • Acts as a chelating agent for removing heavy metals in water
Catalog No. 50-004-06070