Learn More
Apexbio Technology LLC Etifoxine hydrochloride 56776-32-0 10mg
SDP
Supplier:  Apexbio Technology LLC A340110
Encompass_Preferred
Etifoxine hydrochloride (CAS 56776-32-0) is a small molecule that modulates GABAA receptor activity with selectivity towards receptors containing 2 or 3 subunits In neuronal cell culture models it enhances GABAA receptor-mediated responses Etifoxine also binds to the translocator protein (TSPO) a mitochondrial protein involved in neurosteroidogenesis In preclinical and clinical studies etifoxine demonstrates anxiolytic effects without producing sedative muscle relaxant or cognitive impairment typically associated with benzodiazepines Additionally it has been reported to promote axonal regeneration supporting its utility in neuropharmacological and neuroregeneration research
Catalog No. 50-224-0472