Learn More
Medchemexpress LLC Etilefrine hydrochloride | 943-17-9 | 99.9% | C10H16ClNO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYA0144A500MG
Encompass_Preferred
Etilefrine hydrochloride is a sympathetic nerve agonist and AMPK activator that specifically targets α1/β1 adrenergic receptors. It stimulates α1 adrenergic receptors, leading to the contraction of vascular smooth muscle and increased peripheral resistance. Additionally, it enhances myocardial contractility and increases heart rate through β1 receptor stimulation, thereby raising blood pressure and improving cardiac output. It also bidirectionally regulates the AMPK/Akt pathway and modulates phosphorylation levels. It is utilized in cardiovascular research for conditions such as postural hypotension, chylothorax, and improving low cardiac output.
  • Sympathetic nerve agonist
  • AMPK activator
  • Selectively targets α1/β1 adrenergic receptors
  • Stimulates α1 adrenergic receptors
  • Enhances myocardial contractility and heart rate
  • Used in cardiovascular research
  • Regulates AMPK/Akt pathway
Catalog No. 50-004-32476