Learn More
Medchemexpress LLC Etiocholanolone | 53-42-9 | 99.5% | 290.44 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11332010MM/1ML
Encompass_Preferred
Etiocholanolone (5β-Androsterone) is an excreted metabolite of testosterone with anticonvulsant activity. It is a less potent neurosteroid positive allosteric modulator (PAM) of the GABAA receptor than its enantiomer form.
  • Targets GABA receptor and endogenous metabolite
  • Relevant to neurological disease research
  • Exhibits anticonvulsant activity
  • Functions as a less potent neurosteroid positive allosteric modulator (PAM) of the GABAA receptor
Catalog No. 50-004-38627