Learn More
Medchemexpress LLC Etiocholanolone | 53-42-9 | 99.5% | 290.44 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY113320100MG
Encompass_Preferred
Etiocholanolone (5β-Androsterone) is a metabolite of testosterone that exhibits anticonvulsant activity. It functions as a less potent neurosteroid positive allosteric modulator (PAM) of the GABAA receptor compared to its enantiomer form. It is intended for research use only and is not sold to patients.
  • Excreted metabolite of testosterone.
  • Possesses anticonvulsant activity.
  • Acts as a neurosteroid positive allosteric modulator (PAM) of the GABAA receptor.
  • Available in various quantities, including solid and solution forms.
  • Documentation includes Data Sheet, COA, SDS, and Handling Instructions.
  • Solubility information and stock solution preparation guidance are provided.
  • In vivo dissolution protocols are available for different administration routes and animal experiments.
Catalog No. 50-004-38400