Learn More
Medchemexpress LLC Etiocholanolone | 53-42-9 | 99.5% | 290.44 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113320
Encompass_Preferred
Etiocholanolone (5β-Androsterone) is an excreted metabolite of testosterone that exhibits anticonvulsant activity. It functions as a less potent neurosteroid positive allosteric modulator (PAM) of the GABAA receptor compared to its enantiomer form.
  • Excreted metabolite of testosterone.
  • Possesses anticonvulsant activity.
  • Acts as a neurosteroid positive allosteric modulator (PAM) of the GABAA receptor.
Catalog No. 50-004-39304