Learn More
TARGETMOL CHEMICALS INC ETIPREDNOL DICLOACETATE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T317195MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Etiprednol dicloacetate (BNP-166) is a corticosteroid with anti-inflammatory activity.Etiprednol dicloacetate inhibits the accumulation of eosinophils and can be used in the study of inflammatory airway diseases such as asthma and in the treatment of Crohn's disease. purity: 99%
Catalog No. 50-258-0354