Learn More
Medchemexpress LLC Etofenamate | 30544-47-9 | 98.3% | 369.34 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1736110MM/1ML
Encompass_Preferred
Etofenamate is a non-steroid anti-inflammatory agent (NSAID) and a non-selective COX inhibitor. It possesses analgesic, anti-rheumatic, antipyretic, and anti-inflammatory properties. It is used in research for osteoarthritis, arthritis, and other inflammatory diseases.
  • Non-steroid anti-inflammatory agent (NSAID)
  • Non-selective COX inhibitor
  • Possesses analgesic, anti-rheumatic, antipyretic and anti-inflammatory properties.
  • Used in research for osteoarthritis, arthritis and other inflammatory diseases.
Catalog No. 50-003-84214