Learn More
Cayman Chemical Etofenamate-d4 5mg
SDP
Supplier:  Cayman Chemical 307455 MG
Encompass_Preferred
An internal standard for the quantification of etofenamate by GC- or LC-MS
Catalog No. 50-298-9333