Learn More
Selleck Chemical LLC Etomoxir sodium salt 5mg 828934-41-4
SDP
Supplier:  Selleck Chemical LLC S82445MG
Encompass_Preferred
Etomoxir sodium salt is an irreversible inhibitor of carnitine palmitoyltransferase-1 (CPT-1) on the outer face of the inner mitochondrial membrane. Etomoxir enhances palmitate-induced cell apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5842