Learn More
TARGETMOL CHEMICALS INC ETOMOXIR SODIUM SALT 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T453525MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (R)-Etomoxir sodium salt ((R)-Etomoxir sodium salt) is R-form of Etomoxir. Etomoxir is a potent inhibitor of carnitine palmitoyltransferase-I (CPT-1). purity: 99%
Catalog No. 50-257-9871