Learn More
Medchemexpress LLC Etonogestrel | 54048-10-1 | 99.9% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB0652200MG
Encompass_Preferred
Etonogestrel is an active metabolite of Desogestrel, functioning as a third-generation progesterone analogue used in various oral contraceptive preparations. It also serves as a click chemistry reagent, featuring an Alkyne group that enables participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules. Etonogestrel is known to decrease PHOX2B expression by binding to the progesterone nuclear receptor PR-B.
  • Third-generation progesterone analogue
  • Included in oral contraceptive preparations
  • Click chemistry reagent with an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Decreases PHOX2B expression by binding to the progesterone nuclear receptor PR-B
Catalog No. 50-004-08527