Learn More
Medchemexpress LLC Etoricoxib-d4 | 1131345-14-6 | C18H11D4ClN2O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY15321S5MG
Encompass_Preferred
Etoricoxib-d4 (MK-0663-d4) is a deuterium labeled Etoricoxib. It is a non steroidal anti-inflammatory agent, acting as a selective and orally active COX-2 inhibitor, with IC50s of 1.1 μM for COX-2 and 116 μM for COX-1 in human whole blood.
  • Deuterium labeled compound
  • Non steroidal anti-inflammatory agent
  • Selective COX-2 inhibitor
  • Orally active
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-73803