Learn More
Medchemexpress LLC Etoricoxib N1'-oxide | 325855-74-1 | 25mg
SDP
Supplier:  Medchemexpress LLC HYZ470725MG
Encompass_Preferred
Etoricoxib N1 -oxide is an Etoricoxib (HY-15321) metabolite Etoricoxib N1 -oxide does not inhibit COX-1 nor does it significantly inhibit COX-2[1]
Catalog No. 50-002-67063