Learn More
Medchemexpress LLC ETP-45658 | 1198357-79-7 | 99.0% | 311.34 | 1 ML

Supplier: Medchemexpress LLC HY11010910MG
ETP-45658 is a potent PI3K inhibitor, with IC50s of 22.0 nM, 39.8 nM, 129.0 nM and 717.3 nM for PI3Kα, PI3Kδ, PI3Kβ and PI3Kγ, respectively. It also can inhibit DNA-PK (IC50=70.6 nM) and mTOR (IC50=152.0 nM). ETP-45658 can be used for the research of cancer.
- IC50 & Target:
- PI3Kα: 22.0 nM (IC50)
- PI3Kδ: 39.8 nM (IC50)
- PI3Kβ: 129.0 nM (IC50)
- PI3Kγ: 717.3 nM (IC50)
- DNA-PK: 70.6 nM (IC50)
- mTOR: 152.0 nM (IC50)
- In Vitro:
- Decreases in the phosphorylation of FOXO3a, Gsk3-β and p70 S6K in U2OS cells.
- Inhibits the proliferation of MCF7, PC3, 786-O, HTC116, and U251 cells.
- Induces a clear G1 arrest of PC3 cells.
- Inhibits the mutant PI3Kα proteins, H1047R and E545K.
- Induces a dose-dependent increase of GFP-FOXO nuclear translocation in U2foxRELOC cells.
- Decrease the expression of cyclin D1 and p-Akt on serine 473 in U2OS cells.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.