Learn More
Medchemexpress LLC Etravirine-d4 | 1142095-93-9 | 98.3% | C20H11D4BrN6O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY90005S5MG
Encompass_Preferred
Etravirine-d4 is the deuterium-labeled form of Etravirine, which is a non-nucleoside reverse transcriptase inhibitor (NNRTI) used in the treatment of HIV.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32893