Learn More
Medchemexpress LLC EtS-DMAB (HClO-green) | 2929446-76-2 | 99.8% | 223.29 g/mol | C10H13N3OS | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD126510MG
Encompass_Preferred
EtS-DMAB (HClO-green) is a small-molecule fluorescent probe for selective detection and imaging of hypochlorous acid (HOCl) in aqueous solutions and live cells. It exhibits a large Stokes shift with excitation at 440 nm and emission at 610 nm, and is supplied as a viscous liquid with high purity suitable for microscopy and cell-imaging experiments.
  • Selective detection of hypochlorous acid (HOCl) in aqueous solutions and live cells.
  • Large Stokes shift enables clear separation of excitation and emission signals.
  • High purity for reproducible imaging results.
  • Optical properties: excitation 440 nm, emission 610 nm.
  • Store protected from light; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-97992