Learn More
ABclonal Technology ETS1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A156661000UL
Encompass_Preferred
This gene encodes a member of the ETS family of transcription factors, which are defined by the presence of a conserved ETS DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T in target genes. These proteins function either as transcriptional activators or repressors of numerous genes, and are involved in stem cell development, cell senescence and death, and tumorigenesis. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Catalog No. 50-001-06584