Learn More
Molecular Devices Molecular Devices Cardiotoxicity Assay Kit, Fluorescence, Calcium Sensitive, Two-Plate Capacity
Supplier:  Molecular Devices R8210
Encompass_Preferred
The EarlyTox Cardiotoxicity Kit is a fluorescence-based assay for rapid identification of cardiotoxic compounds using stem cell-derived or primary cardiomyocytes. It measures beat frequency, peak amplitude, and beat pattern with a calcium-sensitive indicator and masking technology to improve signal window and reduce background.
  • Provides reagents sufficient for two microplates (96- or 384-well).
  • Uses a calcium-sensitive dye with masking technology to reduce extracellular background.
  • Enables rapid, high-throughput screening of cardiotoxicity in minutes.
  • Includes two vials of component A, one bottle of component B, and three reference compound vials for assay benchmarking.
  • Free of animal-derived components.
  • Compatible with fluorescence plate readers, high-content imagers, and FLIPR-style screening systems.
Catalog No. 50-000-14344