Learn More
Medchemexpress LLC Eudesmin | 526-06-7 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN23571MG
Encompass_Preferred
Eudesmin ((-)-Eudesmin) functions by impairing adipogenic differentiation through the inhibition of the S6K1 signaling pathway. It is noted for its diverse therapeutic properties, which include anti-tumor, anti-inflammatory, and anti-bacterial activities.
  • Impairs adipogenic differentiation through inhibition of the S6K1 signaling pathway.
  • Exhibits diverse therapeutic properties, including anti-tumor, anti-inflammatory, and anti-bacterial activities.
  • Inhibits activation and nuclear translocation of S6K1.
  • Reduces S6K1-mediated phosphorylation of H2B at serine 36 (H2BS36p).
  • Disturbs adipogenesis in mesenchymal stem cells.
Catalog No. 50-004-21351