Learn More
Medchemexpress LLC Eudesmin | 526-06-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN23575MG
Encompass_Preferred
Eudesmin impairs adipogenic differentiation via inhibition of the S6K1 signaling pathway. It possesses diverse therapeutic effects, including anti-tumor, anti-inflammatory, and anti-bacterial activities.
  • Impares adipogenic differentiation via inhibition of the S6K1 signaling pathway.
  • Possesses anti-tumor activities.
  • Possesses anti-inflammatory activities.
  • Possesses anti-bacterial activities.
  • Inhibits activation and nuclear translocation of S6K1.
  • Reduces S6K1-mediated phosphorylation of H2B at serine 36.
Catalog No. 50-004-20255