Learn More
Medchemexpress LLC Eugenol acetate | 93-28-7 | MFCD00026191 | 99.8% | 206.24 g/mol | C12H14O3 | 500 MG

Supplier: Medchemexpress LLC HYW014612500MG
Eugenol acetate (Acetyleugenol) is a phenylpropanoid ester used as a research reagent with reported antibacterial, anti-inflammatory, antioxidant, and anticancer activity. Supplied with batch documentation and safety data, it is intended for biochemical and pharmacological research applications where a well-characterized aromatic ester is required.
- High purity (≈99.8% by HPLC).
- Molecular formula C12H14O3 and molecular weight 206.24 g/mol.
- Useful for antibacterial, anti-inflammatory, antioxidant, and anticancer studies.
- Batch certificate of analysis and safety data sheet available.
- Available in lab pack sizes (for example, 500 mg to 10 g).
- Store at 4°C under inert atmosphere; in solution, store long term at -80°C.
- Intended for research use; follow SDS for handling and disposal.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.