Learn More
Medchemexpress LLC Eugenol-d3 | 1335401-17-6 | 98.5% | 167.22 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0337S50MG
Encompass_Preferred
Eugenol-d3 is the deuterium labeled Eugenol. Eugenol is an essential oil found in cloves with antibacterial, anthelmintic, and antioxidant activity. Eugenol is shown to inhibit lipid peroxidation. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-27212