Learn More
Medchemexpress LLC Eugenol-d3 | 1335401-17-6 | 98.5% | 167.22 g/mol | C10H9D3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0337S10MG
Encompass_Preferred
Eugenol-d3 is the triply deuterated analogue of eugenol, supplied as a high-purity stable isotope standard for analytical and tracer applications. It is intended for use as an internal standard in mass spectrometry and for quantitative metabolic studies.
  • High purity (~98.5%) suitable for analytical workflows.
  • Chemical formula C10H9D3O2 and molecular weight 167.22 g/mol.
  • Used as an internal standard for mass spectrometry and quantitative analysis.
  • Suitable for tracer and metabolic studies requiring stable isotopes.
  • Available in small research quantities (10 mg) for laboratory use.
Catalog No. 50-002-95641