Learn More
Medchemexpress LLC Euk 134 | 81065-76-1 | MFCD08059577 | 98.3% | 416.74 g/mol | C18H18ClMnN2O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10059410MM 1ML
Encompass_Preferred
EUK-134 is a synthetic superoxide dismutase (SOD) and catalase mimetic used as a research antioxidant and mitoprotective agent. It is applied in studies of oxidative stress, ischemia-reperfusion injury, and mitochondrial protection. The compound has molecular formula C18H18ClMnN2O4, molecular weight 416.74 g/mol, and CAS number 81065-76-1. It is commonly supplied as a 10 mM solution in DMSO (1 mL) or as solid quantities for laboratory use.
  • Mimics SOD and catalase activity to scavenge reactive oxygen species.
  • Reduces oxidative damage in cell and tissue models.
  • Mitoprotective properties that support studies of mitochondrial function.
  • Available as 10 mM solution in DMSO, ready for experimental use.
  • High purity suitable for research applications.
  • Storage and handling guidance provided on the safety data sheet.
Catalog No. 50-002-13656