Learn More
Medchemexpress LLC Euk-134 | 81065-76-1 | MFCD08059577 | 98.3% | 416.74 g/mol | C18H18ClMnN2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1005945MG
Encompass_Preferred
EUK-134 is a salen-manganese complex that functions as both a superoxide dismutase and catalase mimetic. It scavenges superoxide and reduces hydrogen peroxide-mediated damage, making it suitable for cell-based assays and in vivo studies of oxidative stress pathways.
  • Combines superoxide dismutase and catalase activities.
  • Scavenges superoxide and reduces hydrogen peroxide-mediated damage.
  • Reported purity of 98.3%.
  • Molecular weight 416.74 g/mol; molecular formula C18H18ClMnN2O4.
  • Available in small research quantities, such as 5 mg.
Catalog No. 50-002-13897