Learn More
Abcam EUK 134, SOD/catalase mimetic, 25MG
Supplier:  Abcam AB14559325MG
Encompass_Preferred
MW 416.7 Da, Purity >98%. SOD/catalase mimetic. Consumes H₂O₂ with an initial reaction rate of 234 μM/min. Ameliorates nNOSμ translocation and skeletal muscle fiber atrophy during short-term mechanical unloading. Shows neuroprotective and anti-ischemic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5129