Learn More
Electron Microscopy Sciences EUKITT 4400 LB Resin 500 ML
SDP
Supplier:  Electron Microscopy Sciences 1532040
Encompass_Preferred
EUKITT 4400 LB is a single-component resin that polymerizes under UV light was specially developed for thin ground sectioning technology. It completely passes through hard and soft tissue and is used for embedding and preparing of sections for medical and technical applications. It is characterized by crystal-clear optics with a favorable refractive index, good fluidity and low shrinkage.
The hardening of the embedding medium is done by UV light of a wavelength of 365 nm with 2 x 9 W. The exposure takes place from two sides. During the exposure within the given time (see instruction manual/operating instructions) temperatures of 50°C will not be exceeded.
Fast staining is possible with all known methods, and with the same high quality.
EUKITT EMS 4400 LB does not contain monomers, therefore the vapour pressure is very low. As a consequence the user is not bothered by intense, toxic smell.
Catalog No. 50-193-0972