Learn More
Apexbio Technology LLC Eupatilin 22368-21-4 20mg
SDP
Supplier:  Apexbio Technology LLC N215920
Encompass_Preferred
Eupatilin (22368-21-4) is a small-molecule agonist targeting peroxisome proliferator-activated receptor alpha (PPAR ) It is designed to modulate PPAR signaling thereby influencing pathways involved in lipid metabolism inflammation suppression and antioxidant response Eupatilin exerts its biological activity primarily through interaction with PPAR receptors modulating downstream signaling cascades associated with inflammation and oxidative stress Based on these pharmacological properties Eupatilin holds research potential in the investigation of inflammatory disorders oxidative damage and metabolic dysregulation
Catalog No. 50-000-90098