Learn More
Medchemexpress LLC Eupatilin | 22368-21-4 | 344.32 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN0783100MG
Encompass_Preferred
Eupatilin is a lipophilic flavonoid isolated from *Artemisia argyi Lévl. et Van.*, functioning as a PPARα agonist and possessing anti-apoptotic, anti-oxidative, and anti-inflammatory properties.
  • PPARα agonist
  • Anti-apoptotic activity
  • Anti-oxidative activity
  • Anti-inflammatory activity
Catalog No. 50-004-17083