Learn More
Medchemexpress LLC Eupatilin | 22368-21-4 | 344.32 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0783
Encompass_Preferred
Eupatilin is a lipophilic flavonoid isolated from Artemisia argyi Lévl. et Van. It acts as a PPARα agonist and exhibits anti-apoptotic, anti-oxidative, and anti-inflammatory activities. It is for research use only and not for sale to patients. This compound appears as a light yellow to yellow solid with the formula C18H16O7 and is classified as a flavonoid.
  • Acts as a PPARα agonist.
  • Exhibits anti-apoptotic, anti-oxidative, and anti-inflammatory activities.
  • Isolated from Artemisia argyi Lévl. et Van.
  • Appears as a light yellow to yellow solid.
  • Has the chemical formula C18H16O7.
  • Classified as a flavonoid.
  • Intended for research use only.
Catalog No. 50-003-89159