Learn More
Medchemexpress LLC Eupatorin | 855-96-9 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN237410MM/1ML
Encompass_Preferred
Eupatorin is an orally active flavonoid with antiproliferative and vasodilatory properties. It downregulates key inflammatory mediators, induces apoptosis and cell cycle arrest, and shows cytotoxic effects against various cancer cell lines. This compound also modulates receptor activities and specific cellular pathways, making it valuable for related research.
  • Orally active flavonoid
  • Antiproliferative and vasodilatory properties
  • Downregulates NF-κB, MMP9, IL-1β, and TNF-α expression
  • Induces apoptosis, G2/M phase cell cycle arrest, and ROS production
  • Exhibits cytotoxic effects on a range of cancer cell lines
  • Useful in research related to breast cancer, hypertension, and leukemia
Catalog No. 50-004-20796