Learn More
Medchemexpress LLC Eupatorin | 855-96-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN23745MG
Encompass_Preferred
Eupatorin is an orally active flavonoid with antiproliferative and vasodilatory properties. It downregulates NF-κB, MMP9, IL-1β, and TNF-α, and induces apoptosis, G2/M phase cell cycle arrest, and reactive oxygen species production. It can be used in research related to breast cancer, hypertension, and leukemia.
  • Orally active flavonoid
  • Antiproliferative and vasodilatory properties
  • Downregulates NF-κB, MMP9, IL-1β, and TNF-α
  • Induces apoptosis and G2/M phase cell cycle arrest
  • Modulates muscarinic and β-adrenergic receptors
  • Inhibits sarcoplasmic reticulum calcium release and calcium channels
  • Activates NO/sGC/cGMP, indomethacin-sensitive, and potassium channel pathways
  • Cytotoxic effects on cancer cell lines
  • Potential research applications in breast cancer, hypertension, and leukemia
Catalog No. 50-004-19344