Learn More
TARGETMOL CHEMICALS INC EUPHORBIASTEROID 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T322025MG
Encompass_Preferred
Also available in 5 mg 10 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Euphorbiasteroid (Euphobiasteroid) (25-50 uM) decreased differentiation of 3T3-L1 cells via reduction of intracellular triglyceride (TG) accumulation. purity: 99%
Catalog No. 50-257-6971