Learn More
Strem, An Ascensus Company CAS# 62667-64-5. 1g. Europium(III) acetate hydrate (99.9%-Eu) (REO). MFCD00150115
SDP
Supplier:  Strem, An Ascensus Company 9363011G
Encompass
CAS# 62667-64-5. 1g. Europium(III) acetate hydrate (99.9%-Eu) (REO). MFCD00150115. Molecular Weight: 329.10. Molecular Formula: Eu(OOCCH3)3∙XH2O. Color/form: white xtl. Strem# 93-6301. http://www.strem.com/catalog/v/93-6301/
Catalog No. 50-901-16021